Product Details

SNP ID
rs146166184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24574749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGGATGGCTCTGCGCGCAGTGA[C/T]GTGTTGCTGGGTGTTTTCCCGTCTC
Phenotype
MIM: 116830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSG PubMed Links

Gene Details

Gene
CTSG
Gene Name
cathepsin G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001911.2 302 Missense Mutation ATC,GTC I89V NP_001902.1
XM_011536499.1 302 Missense Mutation ATC,GTC I103V XP_011534801.1

View Full Product Details