Product Details
- SNP ID
-
rs146341754
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:70046157 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTGACGGAGAAGGCCAGTGTGCC[A/G]GCCGACACGTGGAACTCCTGTCCCT
- Phenotype
-
MIM: 607991
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC8A3
PubMed Links
Gene Details
- Gene
- SLC8A3
- Gene Name
- solute carrier family 8 member A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130417.2 |
1025 |
Silent Mutation |
GCC,GCT |
A229A |
NP_001123889.1 |
NM_033262.4 |
1025 |
Silent Mutation |
GCC,GCT |
A856A |
NP_150287.1 |
NM_058240.3 |
1025 |
Silent Mutation |
GCC,GCT |
A855A |
NP_489479.1 |
NM_182932.2 |
1025 |
Silent Mutation |
GCC,GCT |
A852A |
NP_891977.1 |
NM_182936.2 |
1025 |
Silent Mutation |
GCC,GCT |
A215A |
NP_891981.1 |
NM_183002.2 |
1025 |
Silent Mutation |
GCC,GCT |
A858A |
NP_892114.1 |
XM_006720240.3 |
1025 |
Silent Mutation |
GCC,GCT |
A235A |
XP_006720303.1 |
XM_017021606.1 |
1025 |
Silent Mutation |
GCC,GCT |
A858A |
XP_016877095.1 |
XM_017021607.1 |
1025 |
Silent Mutation |
GCC,GCT |
A857A |
XP_016877096.1 |
XM_017021608.1 |
1025 |
Silent Mutation |
GCC,GCT |
A852A |
XP_016877097.1 |
XM_017021609.1 |
1025 |
Silent Mutation |
GCC,GCT |
A851A |
XP_016877098.1 |
XM_017021610.1 |
1025 |
Intron |
|
|
XP_016877099.1 |
XM_017021611.1 |
1025 |
Silent Mutation |
GCC,GCT |
A242A |
XP_016877100.1 |
View Full Product Details