Product Details

SNP ID
rs146645952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95690793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCAGTGCATACCCGGGAGCTA[C/T]TCTCCTCCGGCCAGATGCCCTTCTC
Phenotype
MIM: 186960 MIM: 603769
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCL1A PubMed Links

Gene Details

Gene
TCL1A
Gene Name
T-cell leukemia/lymphoma 1A
There are no transcripts associated with this gene.

Gene
TCL1B
Gene Name
T-cell leukemia/lymphoma 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004918.3 271 Missense Mutation CTC,TTC L74F NP_004909.1

View Full Product Details