Product Details
- SNP ID
-
rs147118623
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:103100510 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCCTGAATGACGGCCCAGCTAC[C/T]GGCCATTCCCAGGCCACTCCTGAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EXOC3L4
PubMed Links
Gene Details
- Gene
- EXOC3L4
- Gene Name
- exocyst complex component 3 like 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001077594.1 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
NP_001071062.1 |
XM_011537323.2 |
774 |
Silent Mutation |
ACC,ACT |
T155T |
XP_011535625.1 |
XM_011537324.2 |
774 |
Silent Mutation |
ACC,ACT |
T155T |
XP_011535626.1 |
XM_011537325.2 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
XP_011535627.1 |
XM_011537327.2 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
XP_011535629.1 |
XM_011537328.2 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
XP_011535630.1 |
XM_011537329.2 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
XP_011535631.1 |
XM_011537330.2 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
XP_011535632.1 |
XM_011537331.1 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
XP_011535633.1 |
XM_011537332.2 |
774 |
Silent Mutation |
ACC,ACT |
T97T |
XP_011535634.1 |
XM_011537333.2 |
774 |
Silent Mutation |
ACC,ACT |
T134T |
XP_011535635.1 |
XM_011537334.1 |
774 |
Intron |
|
|
XP_011535636.1 |
View Full Product Details