Product Details

SNP ID
rs147469794
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73713031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGCGGGGCCTCTAAGACTCTC[A/C]ATCAACCGCCGCCTCTTTTCTACAT
Phenotype
MIM: 610062 MIM: 604010
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DNAL1 PubMed Links

Gene Details

Gene
DNAL1
Gene Name
dynein axonemal light chain 1
There are no transcripts associated with this gene.

Gene
ELMSAN1
Gene Name
ELM2 and Myb/SANT domain containing 1
There are no transcripts associated with this gene.

Gene
PNMA1
Gene Name
paraneoplastic Ma antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006029.4 1395 Missense Mutation ATG,ATT M203I NP_006020.4

View Full Product Details