Product Details
- SNP ID
-
rs147627228
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:35403288 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCCTCGAAAGTCTCGGAGCTCAG[A/G]ATCACAGCCAGCTCCCAGAAGTGCC
- Phenotype
-
MIM: 164008
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NFKBIA
PubMed Links
Gene Details
- Gene
- NFKBIA
- Gene Name
- NFKB inhibitor alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020529.2 |
519 |
Missense Mutation |
CCT,TCT |
P137S |
NP_065390.1 |
View Full Product Details