Product Details

SNP ID
rs148009150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:30567924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATATTTTGTGTGATTTCAGTCTT[A/G]TAAAATGTATTATTTTGTGGTCTAA
Phenotype
MIM: 611299
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G2E3 PubMed Links

Gene Details

Gene
G2E3
Gene Name
G2/M-phase specific E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308097.1 Intron NP_001295026.1
NM_017769.4 Intron NP_060239.2
XM_011536925.2 Intron XP_011535227.1
XM_011536926.1 Intron XP_011535228.1

View Full Product Details