Product Details

SNP ID
rs148039355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93718448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATGGCTTGTTGGTGGATAAGAC[A/G]GCATCAACTTGTACATTTGCCCATT
Phenotype
MIM: 613851 MIM: 616884
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRIMA1 PubMed Links

Gene Details

Gene
PRIMA1
Gene Name
proline rich membrane anchor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178013.3 3500 UTR 3 NP_821092.1
XM_011536456.2 3500 UTR 3 XP_011534758.1
Gene
UNC79
Gene Name
unc-79 homolog (C. elegans)
There are no transcripts associated with this gene.

View Full Product Details