Product Details

SNP ID
rs148053975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104594011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCAGCAGCTCCGGCCTGTCG[A/G]CTCTAAGGAGGGCGGGTAAACAGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf180 PubMed Links

Gene Details

Gene
C14orf180
Gene Name
chromosome 14 open reading frame 180
There are no transcripts associated with this gene.

Gene
TMEM179
Gene Name
transmembrane protein 179
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286389.1 Intron NP_001273318.1
NM_001286390.1 Intron NP_001273319.1
XM_011536745.2 Intron XP_011535047.1
XM_011536746.2 Intron XP_011535048.1
XM_011536750.2 Intron XP_011535052.1

View Full Product Details