Product Details

SNP ID
rs148451324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20782000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAGAGCTCAAAGCCTGTCAAC[A/G]TGACTGACTGCAGACTCACTTCAGG
Phenotype
MIM: 601981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNASE6 PubMed Links

Gene Details

Gene
RNASE6
Gene Name
ribonuclease A family member k6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005615.4 594 Missense Mutation ATG,GTG M101V NP_005606.1
XM_017021566.1 594 Missense Mutation ATG,GTG M101V XP_016877055.1
XM_017021567.1 594 Missense Mutation ATG,GTG M101V XP_016877056.1

View Full Product Details