Product Details

SNP ID
rs148636133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104727489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGCTGAGTCTCCTCTGAGCCA[A/G]GTCTGCCCTCCCTGCTCTGCGGCCA
Phenotype
MIM: 612498 MIM: 610982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADSSL1 PubMed Links

Gene Details

Gene
ADSSL1
Gene Name
adenylosuccinate synthase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320424.1 Intron NP_001307353.1
NM_152328.4 Intron NP_689541.1
NM_199165.2 Intron NP_954634.1
XM_006720026.3 Intron XP_006720089.1
XM_011536412.1 Intron XP_011534714.1
XM_011536414.2 Intron XP_011534716.1
Gene
INF2
Gene Name
inverted formin, FH2 and WH2 domain containing
There are no transcripts associated with this gene.

Gene
LOC388022
Gene Name
uncharacterized LOC388022
There are no transcripts associated with this gene.

View Full Product Details