Product Details

SNP ID
rs148655599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20034195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTAGGATGTAGGTGTCCTTGCA[G/T]GCAAGTTTAATCACAAGGGGAAGGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K13 PubMed Links

Gene Details

Gene
OR4K13
Gene Name
olfactory receptor family 4 subfamily K member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004714.1 564 Silent Mutation GCA,GCC A188A NP_001004714.1

View Full Product Details