Product Details

SNP ID
rs148766492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105051281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACACCACAGAGGCTGTGTACA[A/G]CCTTTCCTCCAAGCCGCACATGGCG
Phenotype
MIM: 606167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR132 PubMed Links

Gene Details

Gene
GPR132
Gene Name
G protein-coupled receptor 132
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278694.1 994 Silent Mutation NP_001265623.1
NM_001278695.1 994 Silent Mutation NP_001265624.1
NM_001278696.1 994 Silent Mutation NP_001265625.1
NM_013345.3 994 Silent Mutation NP_037477.1

View Full Product Details