Product Details

SNP ID
rs148786753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75279946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGTCACTGCCATCTCGACCAGT[A/C]CGGACCTGCAGTGGCTGGTGCAGCC
Phenotype
MIM: 164810
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FOS PubMed Links

Gene Details

Gene
FOS
Gene Name
Fos proto-oncogene, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005252.3 416 Missense Mutation ACG,CCG T71P NP_005243.1

View Full Product Details