Product Details

SNP ID
rs149841422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23633068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCTCTGGGCTAACATGAATGT[G/T]GAGCTGGGAAATTAGGATTTGGGTG
Phenotype
MIM: 615194
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DHRS2 PubMed Links

Gene Details

Gene
DHRS2
Gene Name
dehydrogenase/reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318835.1 Intron NP_001305764.1
NM_005794.3 Intron NP_005785.1
NM_182908.4 Intron NP_878912.1
XM_005267249.1 Intron XP_005267306.1
XM_006720001.3 Intron XP_006720064.1
XM_011536338.1 Intron XP_011534640.1
XM_011536339.2 Intron XP_011534641.1

View Full Product Details