Product Details

SNP ID
rs149855008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93721539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGCCATTTTCGTCTTTTCTCA[G/T]TGGTTTCCTGGAAGTGGGGGGAGGG
Phenotype
MIM: 613851
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRIMA1 PubMed Links

Gene Details

Gene
PRIMA1
Gene Name
proline rich membrane anchor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178013.3 409 Missense Mutation ATG,CTG M123L NP_821092.1
XM_011536456.2 409 Missense Mutation ATG,CTG M123L XP_011534758.1

View Full Product Details