Product Details

SNP ID
rs149973422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20956204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATGTGACAACAGAGATCAACGA[A/C]GAGACCCTCCACAGTATCCGGTGGT
Phenotype
MIM: 131410
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNASE2 PubMed Links

Gene Details

Gene
RNASE2
Gene Name
ribonuclease A family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002934.2 504 Silent Mutation AGA,CGA R145R NP_002925.1

View Full Product Details