Product Details

SNP ID
rs149983760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52006619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGTTGTTTTGAATGACACGCC[A/G]TCCAGTTCCATCAGGTAGTGTACAC
Phenotype
MIM: 610858 MIM: 605399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf166 PubMed Links

Gene Details

Gene
C14orf166
Gene Name
chromosome 14 open reading frame 166
There are no transcripts associated with this gene.

Gene
NID2
Gene Name
nidogen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007361.3 4156 Missense Mutation CGG,TGG R1308W NP_031387.3
XM_005267405.4 4156 Missense Mutation CGG,TGG R1335W XP_005267462.1
XM_005267406.4 4156 Missense Mutation CGG,TGG R1287W XP_005267463.1
XM_005267407.4 4156 Missense Mutation CGG,TGG R1260W XP_005267464.1

View Full Product Details