Product Details

SNP ID
rs150018148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:29577358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTACTGCAGCCCCTGCTCGCC[C/T]GCATACTTCTCCCACCTCAGGTCAT
Phenotype
MIM: 605435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKD1 PubMed Links

Gene Details

Gene
PRKD1
Gene Name
protein kinase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002742.2 5180 Silent Mutation GCA,GCG A873A NP_002733.2
XM_005267859.1 5180 Silent Mutation GCA,GCG A881A XP_005267916.1
XM_011536964.2 5180 Silent Mutation GCA,GCG A805A XP_011535266.1
XM_011536965.2 5180 Silent Mutation GCA,GCG A785A XP_011535267.1
XM_017021461.1 5180 Silent Mutation GCA,GCG A785A XP_016876950.1
XM_017021462.1 5180 Silent Mutation GCA,GCG A708A XP_016876951.1

View Full Product Details