Product Details

SNP ID
rs150213507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60247094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCACTCTAAATGGTTGCCATAT[A/G]AGCTTTTGTAGTTCAGAATGTTAAT
Phenotype
MIM: 606108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927702 PubMed Links

Gene Details

Gene
LOC101927702
Gene Name
uncharacterized LOC101927702
There are no transcripts associated with this gene.

Gene
PPM1A
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021003.4 Intron NP_066283.1
NM_177951.2 Intron NP_808820.1
NM_177952.2 Intron NP_808821.2
XM_005267779.1 Intron XP_005267836.1
XM_005267781.1 Intron XP_005267838.1
XM_011536872.1 Intron XP_011535174.1
XM_011536876.1 Intron XP_011535178.1
XM_011536877.1 Intron XP_011535179.1
XM_011536878.1 Intron XP_011535180.1
XM_011536879.2 Intron XP_011535181.1
XM_011536880.2 Intron XP_011535182.1
XM_011536881.2 Intron XP_011535183.1
XM_011536882.2 Intron XP_011535184.1
XM_011536883.1 Intron XP_011535185.1
XM_011536884.1 Intron XP_011535186.1
XM_017021381.1 Intron XP_016876870.1
XM_017021382.1 Intron XP_016876871.1
XM_017021383.1 Intron XP_016876872.1
XM_017021384.1 Intron XP_016876873.1
XM_017021385.1 Intron XP_016876874.1
XM_017021386.1 Intron XP_016876875.1
XM_017021387.1 Intron XP_016876876.1

View Full Product Details