Product Details

SNP ID
rs150328141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92797758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGCATCTGTTCCTAGGCCTTC[A/G]TCCCATTTTGTGCGAAGAAAAAAGT
Phenotype
MIM: 606918
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLGA5 PubMed Links

Gene Details

Gene
GOLGA5
Gene Name
golgin A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005113.3 Intron NP_005104.3
XM_011537420.2 Intron XP_011535722.1

View Full Product Details