Product Details

SNP ID
rs150451114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59596047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAAGTGCAAATATCCAGAGTCC[A/G]TAATTGAATCCATTAGGAACTACAG
Phenotype
MIM: 600865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTN1 PubMed Links

Gene Details

Gene
RTN1
Gene Name
reticulon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021136.2 1633 UTR 3 NP_066959.1
NM_206852.2 1633 UTR 3 NP_996734.1
XM_011537063.2 1633 Intron XP_011535365.1
XM_017021578.1 1633 UTR 3 XP_016877067.1

View Full Product Details