Product Details

SNP ID
rs150553508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104938833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCCTCCTGAGTCCTAGAGTGTT[A/G]AGTCATTGTTGTGTACACTCTAGCC
Phenotype
MIM: 608570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHNAK2 PubMed Links

Gene Details

Gene
AHNAK2
Gene Name
AHNAK nucleoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138420.2 16738 Nonsense Mutation CAA,TAA Q5540* NP_612429.2
XM_005267299.1 16738 Nonsense Mutation CAA,TAA Q5440* XP_005267356.1
Gene
PLD4
Gene Name
phospholipase D family member 4
There are no transcripts associated with this gene.

View Full Product Details