Product Details

SNP ID
rs150583863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20556478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGCGATATGACAAAGACACCAT[C/T]CTCACTGAGGAAACTTCTGTGTTCA
Phenotype
MIM: 614014
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASE9 PubMed Links

Gene Details

Gene
RNASE9
Gene Name
ribonuclease A family member 9 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001673.3 752 Missense Mutation GAT,TAT D198Y NP_001001673.2
NM_001110356.1 752 Missense Mutation GAT,TAT D198Y NP_001103826.1
NM_001110357.1 752 Missense Mutation GAT,TAT D198Y NP_001103827.1
NM_001110358.1 752 Missense Mutation GAT,TAT D203Y NP_001103828.1
NM_001110359.1 752 Missense Mutation GAT,TAT D203Y NP_001103829.1
NM_001110360.1 752 Missense Mutation GAT,TAT D203Y NP_001103830.1
NM_001110361.1 752 Missense Mutation GAT,TAT D203Y NP_001103831.1
NM_001289110.1 752 Missense Mutation GAT,TAT D198Y NP_001276039.1

View Full Product Details