Product Details

SNP ID
rs151072003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74853414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTAAAACTTAGGTTTGGCAGAT[G/T]CCTTCATTTTTGTGAAAATAGGGCA
Phenotype
MIM: 615094
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PROX2 PubMed Links

Gene Details

Gene
PROX2
Gene Name
prospero homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080408.2 2816 UTR 3 NP_001073877.2
NM_001243007.1 2816 UTR 3 NP_001229936.1
XM_005267543.3 2816 Intron XP_005267600.1
XM_017021222.1 2816 Intron XP_016876711.1

View Full Product Details