Product Details

SNP ID
rs151240998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44505644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAGTAGGAGCCTCTTCTGCAGG[G/T]GTCTCCATAGCTGTTGAAAGCTGAA
Phenotype
MIM: 611779
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FSCB PubMed Links

Gene Details

Gene
FSCB
Gene Name
fibrous sheath CABYR binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032135.3 1653 Silent Mutation ACA,ACC T448T NP_115511.3
Gene
LOC105370473
Gene Name
uncharacterized LOC105370473
There are no transcripts associated with this gene.

View Full Product Details