Product Details

SNP ID
rs111784082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65154793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAAATCTAACCTTATCCATGC[C/T]GAATAAGGCCTGGTACTGAGGAATA
Phenotype
MIM: 615611
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLPX PubMed Links

Gene Details

Gene
CLPX
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006660.4 1867 Missense Mutation AGC,GGC S534G NP_006651.2
XM_011521164.2 1867 Intron XP_011519466.1

View Full Product Details