Product Details

SNP ID
rs111882402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:71825260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACTAATTCCCTCAACCCAGGGA[C/T]GAATGGCTCTCATCCCCCTGTCCAG
Phenotype
MIM: 604875 MIM: 604485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO9A PubMed Links

Gene Details

Gene
MYO9A
Gene Name
myosin IXA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006901.3 9414 Intron NP_008832.2
XM_006720539.2 9414 UTR 3 XP_006720602.1
XM_011521613.2 9414 UTR 3 XP_011519915.1
XM_011521614.2 9414 UTR 3 XP_011519916.1
XM_011521615.2 9414 UTR 3 XP_011519917.1
XM_011521616.2 9414 UTR 3 XP_011519918.1
XM_011521617.2 9414 UTR 3 XP_011519919.1
XM_011521618.2 9414 UTR 3 XP_011519920.1
XM_011521619.2 9414 UTR 3 XP_011519921.1
XM_011521620.2 9414 UTR 3 XP_011519922.1
XM_011521621.2 9414 UTR 3 XP_011519923.1
XM_011521622.2 9414 UTR 3 XP_011519924.1
XM_011521623.2 9414 UTR 3 XP_011519925.1
XM_017022228.1 9414 UTR 3 XP_016877717.1
XM_017022229.1 9414 UTR 3 XP_016877718.1
XM_017022230.1 9414 UTR 3 XP_016877719.1
Gene
NR2E3
Gene Name
nuclear receptor subfamily 2 group E member 3
There are no transcripts associated with this gene.

View Full Product Details