Product Details

SNP ID
rs111962818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:33319068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGTAAAGGCATTGCACCCGAC[C/T]AGTGACAGGTACTGGCATCAACTCT
Phenotype
MIM: 180903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928134 PubMed Links

Gene Details

Gene
LOC101928134
Gene Name
uncharacterized LOC101928134
There are no transcripts associated with this gene.

Gene
RYR3
Gene Name
ryanodine receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001036.4 Intron NP_001027.3
NM_001243996.2 Intron NP_001230925.1
XM_011521880.2 Intron XP_011520182.1
XM_017022468.1 Intron XP_016877957.1
XM_017022469.1 Intron XP_016877958.1
XM_017022470.1 Intron XP_016877959.1
XM_017022471.1 Intron XP_016877960.1
XM_017022472.1 Intron XP_016877961.1
XM_017022473.1 Intron XP_016877962.1
XM_017022474.1 Intron XP_016877963.1
XM_017022475.1 Intron XP_016877964.1
XM_017022476.1 Intron XP_016877965.1
XM_017022477.1 Intron XP_016877966.1

View Full Product Details