Product Details

SNP ID
rs112313613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41561250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGGGTGAAGGATGGGGCTGTGG[C/T]CCAGAACTTGGACCAGTTGTACATC
Phenotype
MIM: 600341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TYRO3 PubMed Links

Gene Details

Gene
TYRO3
Gene Name
TYRO3 protein tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006293.3 285 Missense Mutation GCC,GTC A83V NP_006284.2
XM_017022542.1 285 Missense Mutation GCC,GTC A38V XP_016878031.1
XM_017022543.1 285 Missense Mutation GCC,GTC A83V XP_016878032.1

View Full Product Details