Product Details

SNP ID
rs112535272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45366156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTAAAGGAGATGATATGCACTC[C/T]GTAGTCTGGAGCAAGATGCCTACGC
Phenotype
MIM: 602360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATM PubMed Links

Gene Details

Gene
GATM
Gene Name
glycine amidinotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321015.1 1197 Missense Mutation AGA,GGA R161G NP_001307944.1
NM_001482.2 1197 Missense Mutation AGA,GGA R290G NP_001473.1

View Full Product Details