Product Details

SNP ID
rs113181643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64166641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAAATGGCTCATCTTTCCTGA[A/G]ACATTCCTAATAGACTGATAAGCCT
Phenotype
MIM: 606274 MIM: 123841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSNK1G1 PubMed Links

Gene Details

Gene
CSNK1G1
Gene Name
casein kinase 1 gamma 1
There are no transcripts associated with this gene.

Gene
PPIB
Gene Name
peptidylprolyl isomerase B
There are no transcripts associated with this gene.

Gene
SNX22
Gene Name
sorting nexin 22
There are no transcripts associated with this gene.

View Full Product Details