Product Details

SNP ID
rs113294093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65150871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGGCCATCCTTCTTCTTCAAC[A/T]CCAGAGTCATACTCCTCTTCAGAGG
Phenotype
MIM: 615611
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLPX PubMed Links

Gene Details

Gene
CLPX
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006660.4 2121 Silent Mutation GGA,GGT G618G NP_006651.2
XM_011521164.2 2121 Intron XP_011519466.1

View Full Product Details