Product Details

SNP ID
rs113509489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101806323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTGGGATTCATGGTTACTGCC[A/G]ATAGTGGATTTATTTCTCTGGCTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371030 PubMed Links

Gene Details

Gene
LOC105371030
Gene Name
uncharacterized LOC105371030
There are no transcripts associated with this gene.

Gene
OR4F6
Gene Name
olfactory receptor family 4 subfamily F member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005326.1 604 Missense Mutation AAT,GAT N202D NP_001005326.1

View Full Product Details