Product Details
- SNP ID
-
rs113686035
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:34981384 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACATAACCGAATCTAAGGCAACC[A/G]CTTTCCCTTGTGGTCGTTCTTTAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF770
PubMed Links
Gene Details
- Gene
- ZNF770
- Gene Name
- zinc finger protein 770
View Full Product Details