Product Details

SNP ID
rs114178009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65445993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTTAAAATTCTAGAATATTTT[A/G]ATAAGGAAATACCAAATGGAAACTG
Phenotype
MIM: 606819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPP8 PubMed Links

Gene Details

Gene
DPP8
Gene Name
dipeptidyl peptidase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320875.1 2461 UTR 3 NP_001307804.1
NM_001320876.1 2461 UTR 3 NP_001307805.1
NM_017743.5 2461 UTR 3 NP_060213.2
NM_130434.4 2461 UTR 3 NP_569118.1
NM_197960.3 2461 UTR 3 NP_932064.1
NM_197961.3 2461 UTR 3 NP_932065.1
XM_005254505.1 2461 UTR 3 XP_005254562.1
XM_011521730.1 2461 UTR 3 XP_011520032.1
XM_011521731.1 2461 UTR 3 XP_011520033.1
XM_011521732.2 2461 UTR 3 XP_011520034.1
XM_011521733.1 2461 UTR 3 XP_011520035.1
XM_011521734.1 2461 UTR 3 XP_011520036.1
XM_017022373.1 2461 UTR 3 XP_016877862.1
XM_017022374.1 2461 UTR 3 XP_016877863.1
XM_017022375.1 2461 UTR 3 XP_016877864.1
XM_017022376.1 2461 UTR 3 XP_016877865.1
XM_017022377.1 2461 UTR 3 XP_016877866.1
XM_017022378.1 2461 UTR 3 XP_016877867.1
XM_017022379.1 2461 UTR 3 XP_016877868.1
XM_017022380.1 2461 Intron XP_016877869.1
XM_017022381.1 2461 UTR 3 XP_016877870.1
XM_017022382.1 2461 UTR 3 XP_016877871.1
XM_017022383.1 2461 UTR 3 XP_016877872.1

View Full Product Details