Product Details

SNP ID
rs114319739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78107723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTATGGGGTCCAAGACCGGGC[A/G]GATGTTGGGTGGAATCTGAGAAGCT
Phenotype
MIM: 605564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIB2 PubMed Links

Gene Details

Gene
CIB2
Gene Name
calcium and integrin binding family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271888.1 Intron NP_001258817.1
NM_001271889.1 Intron NP_001258818.1
NM_001301224.1 Intron NP_001288153.1
NM_006383.3 Intron NP_006374.1
XM_005254126.3 Intron XP_005254183.1
XM_006720374.2 Intron XP_006720437.1
XM_011521161.2 Intron XP_011519463.1
Gene
SH2D7
Gene Name
SH2 domain containing 7
There are no transcripts associated with this gene.

View Full Product Details