Product Details

SNP ID
rs114754379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76935619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAATATGATAAAAACAGTGATG[A/G]TACTGTGACTTGGGATGAATATAAC
Phenotype
MIM: 602584
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RCN2 PubMed Links

Gene Details

Gene
RCN2
Gene Name
reticulocalbin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271837.1 565 Missense Mutation GAT,GGT D115G NP_001258766.1
NM_002902.2 565 Missense Mutation GAT,GGT D115G NP_002893.1

View Full Product Details