Product Details

SNP ID
rs114823813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:77052849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGTCTCTCTGCAGCAGCTAAG[A/G]GGGACACTCTGGTTTTTGGTTTCTT
Phenotype
MIM: 613134
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN3 PubMed Links

Gene Details

Gene
TSPAN3
Gene Name
tetraspanin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168412.1 787 Silent Mutation CCC,CCT P107P NP_001161884.1
NM_005724.5 787 Silent Mutation CCC,CCT P171P NP_005715.1
NM_198902.2 787 Silent Mutation CCC,CCT P146P NP_944492.1
XM_017021857.1 787 Silent Mutation CCC,CCT P132P XP_016877346.1

View Full Product Details