Product Details

SNP ID
rs114868337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40765178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCTTATTTCTTAAAAGATATGG[C/T]TTTATTGATATGTAATGTGCATACC
Phenotype
MIM: 616844 MIM: 602437 MIM: 611873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf62 PubMed Links

Gene Details

Gene
C15orf62
Gene Name
chromosome 15 open reading frame 62
There are no transcripts associated with this gene.

Gene
DNAJC17
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
There are no transcripts associated with this gene.

Gene
GCHFR
Gene Name
GTP cyclohydrolase I feedback regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005258.2 Intron NP_005249.1
Gene
RMDN3
Gene Name
regulator of microtubule dynamics 3
There are no transcripts associated with this gene.

View Full Product Details