Product Details

SNP ID
rs114917692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:71113653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTACCACCTACTGGTATGGTTT[C/T]CTGACTATATTTCATCTTCTTCGCC
Phenotype
MIM: 614476
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CT62 PubMed Links

Gene Details

Gene
CT62
Gene Name
cancer/testis antigen 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102658.1 739 UTR 5 NP_001096128.1
XM_006720429.3 739 UTR 5 XP_006720492.1
Gene
THSD4
Gene Name
thrombospondin type 1 domain containing 4
There are no transcripts associated with this gene.

View Full Product Details