Product Details

SNP ID
rs115055554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45028789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGACCTAAGCATTTTTAAATACT[C/T]TTATTTATTTTTAAAAATAAGATAA
Phenotype
MIM: 182500
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SORD PubMed Links

Gene Details

Gene
SORD
Gene Name
sorbitol dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003104.5 Intron NP_003095.2

View Full Product Details