Product Details

SNP ID
rs115389253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84817021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCCCTGGCCCCGGCCAGGCCT[C/G]GTGGACCCCTCATATGCCACACGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALPK3 PubMed Links

Gene Details

Gene
ALPK3
Gene Name
alpha kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020778.4 342 Missense Mutation CGT,GGT R59G NP_065829.3
XM_017022444.1 342 Missense Mutation CGT,GGT R59G XP_016877933.1
XM_017022445.1 342 Missense Mutation CGT,GGT R59G XP_016877934.1

View Full Product Details