Product Details

SNP ID
rs115767399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:97963882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTTTTAGGAGCACTGGGATT[A/G]CCTTGAGCCTTTCACTCCACAGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARRDC4 PubMed Links

Gene Details

Gene
ARRDC4
Gene Name
arrestin domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183376.2 Intron NP_899232.2

View Full Product Details