Product Details

SNP ID
rs116034611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34789408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCACATACATCTCCTTAAAGCC[C/T]CCAGGCCACACGCACTGGGGGATTA
Phenotype
MIM: 102540
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTC1 PubMed Links

Gene Details

Gene
ACTC1
Gene Name
actin, alpha, cardiac muscle 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005159.4 2381 UTR 3 NP_005150.1
Gene
LOC101928174
Gene Name
uncharacterized LOC101928174
There are no transcripts associated with this gene.

View Full Product Details