Product Details

SNP ID
rs117117636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:94242457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGTAAGTGCCTGCCTACGCTG[C/T]TATGTCAGCTTTCTCAGAGATCAGG
Phenotype
MIM: 616297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCTP2 PubMed Links

Gene Details

Gene
MCTP2
Gene Name
multiple C2 and transmembrane domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159643.1 Intron NP_001153115.1
NM_001159644.1 Intron NP_001153116.1
NM_018349.3 Intron NP_060819.3
XM_005254955.3 Intron XP_005255012.1
XM_005254960.2 Intron XP_005255017.1
XM_006720603.2 Intron XP_006720666.1
XM_011521770.1 Intron XP_011520072.1
XM_011521771.2 Intron XP_011520073.1
XM_011521772.2 Intron XP_011520074.1
XM_011521773.2 Intron XP_011520075.1
XM_011521774.2 Intron XP_011520076.1
XM_011521775.2 Intron XP_011520077.1
XM_017022403.1 Intron XP_016877892.1
XM_017022404.1 Intron XP_016877893.1
XM_017022405.1 Intron XP_016877894.1

View Full Product Details