Product Details

SNP ID
rs117481368
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64462387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTTCTACTCTTAGTTGAATCAT[C/T]ATATTTAATTCTTTTATATGAATGG
Phenotype
MIM: 604501
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TRIP4 PubMed Links

Gene Details

Gene
TRIP4
Gene Name
thyroid hormone receptor interactor 4
There are no transcripts associated with this gene.

Gene
ZNF609
Gene Name
zinc finger protein 609
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015042.1 Intron NP_055857.1
XM_011521386.2 Intron XP_011519688.1
XM_017022021.1 Intron XP_016877510.1

View Full Product Details