Product Details

SNP ID
rs117551580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101818591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAACCTCTCCACTACCTGACCAT[C/G]ATGAGCCCAAGAATGTGTCTATACT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4F15 PubMed Links

Gene Details

Gene
OR4F15
Gene Name
olfactory receptor family 4 subfamily F member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001674.1 405 Missense Mutation ATC,ATG I135M NP_001001674.1

View Full Product Details