Product Details

SNP ID
rs117579924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99137602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTCAATCCCTATTAGCCTCCT[A/G]TAAGAAAGGACTTCCTGAAGTTCAC
Phenotype
MIM: 606087
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNM PubMed Links

Gene Details

Gene
SYNM
Gene Name
synemin
There are no transcripts associated with this gene.

Gene
TTC23
Gene Name
tetratricopeptide repeat domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288615.1 2144 UTR 3 NP_001275544.1
NM_001288616.1 2144 UTR 3 NP_001275545.1
XM_011521927.2 2144 UTR 3 XP_011520229.1
XM_011521929.1 2144 UTR 3 XP_011520231.1
XM_011521930.1 2144 UTR 3 XP_011520232.1
XM_011521931.1 2144 UTR 3 XP_011520233.1
XM_011521932.1 2144 UTR 3 XP_011520234.1
XM_011521933.1 2144 UTR 3 XP_011520235.1
XM_011521934.1 2144 UTR 3 XP_011520236.1
XM_011521935.1 2144 UTR 3 XP_011520237.1
XM_011521936.1 2144 UTR 3 XP_011520238.1
XM_011521937.1 2144 UTR 3 XP_011520239.1
XM_011521939.1 2144 UTR 3 XP_011520241.1
XM_011521940.2 2144 UTR 3 XP_011520242.1
XM_011521941.2 2144 UTR 3 XP_011520243.1
XM_011521943.2 2144 Intron XP_011520245.1
XM_011521944.2 2144 Intron XP_011520246.1
XM_011521945.2 2144 Intron XP_011520247.1
XM_017022509.1 2144 UTR 3 XP_016877998.1
XM_017022510.1 2144 UTR 3 XP_016877999.1
XM_017022511.1 2144 UTR 3 XP_016878000.1
XM_017022512.1 2144 UTR 3 XP_016878001.1
XM_017022513.1 2144 UTR 3 XP_016878002.1
XM_017022514.1 2144 UTR 3 XP_016878003.1
XM_017022515.1 2144 UTR 3 XP_016878004.1
XM_017022516.1 2144 UTR 3 XP_016878005.1
XM_017022517.1 2144 UTR 3 XP_016878006.1
XM_017022518.1 2144 UTR 3 XP_016878007.1

View Full Product Details