Product Details

SNP ID
rs118068328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74830178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATGGCAGGTGGAGACGTGGAG[C/T]TGCCCCGGGAGCTGGCCAAGATGAT
Phenotype
MIM: 609585 MIM: 609548 MIM: 613472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPLX3 PubMed Links

Gene Details

Gene
CPLX3
Gene Name
complexin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030005.2 395 Silent Mutation CTG,TTG L101L NP_001025176.1
Gene
LMAN1L
Gene Name
lectin, mannose binding 1 like
There are no transcripts associated with this gene.

Gene
ULK3
Gene Name
unc-51 like kinase 3
There are no transcripts associated with this gene.

View Full Product Details